2-(Chloromethyl)-4-(trifluoromethyl)pyridine 95+%

Leverantör: Apollo Scientific
Varning

PC200260-250MG PC200260-1G
APOSPC200260-250MGEA 3075 SEK
APOSPC200260-250MG APOSPC200260-1G
2-(Chloromethyl)-4-(trifluoromethyl)pyridine 95+%
2-(Chloromethyl)-4-(trifluoromethyl)pyridine
Formel: C₇H₅ClF₃N
Kokpunkt: 199 °C
Storage Temperature: Omgivande
MDL Number: MFCD10697599
CAS nummer: 215867-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR