4-Chloro-2-(difluoromethyl)pyridine hydrochloride
Leverantör: Apollo Scientific
Varning
Synonyms: 4-Chloro-2-(difluoromethyl)pyridine
PC200123-500MG PC200123-1G PC200123-5G
APOSPC200123-500MGEA 3875 SEK
APOSPC200123-500MG APOSPC200123-1G APOSPC200123-5G
4-Chloro-2-(difluoromethyl)pyridine hydrochloride
4-Chloro-2-(difluoromethyl)pyridine hydrochloride
Formel: C₆H₅Cl₂F₂N
MW: 200,02 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD23701768
CAS nummer: 1193104-11-8