4-Bromo-1-ethyl-2-fluorobenzene

Leverantör: Apollo Scientific
Varning

PC200008-1G PC200008-500MG
APOSPC200008-1GEA 4875 SEK
APOSPC200008-1G APOSPC200008-500MG
4-Bromo-1-ethyl-2-fluorobenzene
4-Bromo-1-ethyl-2-fluorobenzene
Formel: C₈H₈BrF
Storage Temperature: Omgivande
MDL Number: MFCD16619427
CAS nummer: 627463-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR