Allyl-2,2,3,3,3-pentafluoropropyl ether 97%
Leverantör: Apollo Scientific
PC1975-1G PC1975-25G PC1975-5G
APOSPC1975-1GEA 930 SEK
APOSPC1975-1G APOSPC1975-25G APOSPC1975-5G
Allyl-2,2,3,3,3-pentafluoropropyl ether 97%
Allyl-2,2,3,3,3-pentafluoropropyl ether
Formel: C₆H₇F₅O
MW: 190.11 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD02093320
CAS nummer: 186907-75-5