Methyl perfluoro-3,6-dioxaheptanoate
Leverantör: Apollo Scientific
PC1947-25G PC1947-5G PC1947-100G
APOSPC1947-25GEA 1800 SEK
APOSPC1947-25G APOSPC1947-5G APOSPC1947-100G
Methyl perfluoro-3,6-dioxaheptanoate
Methyl perfluoro-3,6-dioxaheptanoate
Formel: C₆H₃F₉O₄
Storage Temperature: Omgivande
MDL Number: MFCD03094140
CAS nummer: 39187-41-2