3-Chlor-4-fluorothiophenol, Technical Grade
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
1-Chloro-2-fluoro-5-mercaptobenzene, 3-Chloro-4-fluorobenzenethiol, 3-Chlor-4-fluorobenzenethiol, 3-Chlor-4-fluorobenzene-1-thiol
PC1889M-10G
PC1889M-1G
APOSPC1889M-10GEA
4275
SEK
APOSPC1889M-10G
APOSPC1889M-1G
3-Chlor-4-fluorothiophenol, Technical Grade
3-Chlor-4-fluorothiophenol
Formel:
C₆H₄ClFS MW: 162.61 g/mol Kokpunkt: 68…70 °C/2mmHg Storage Temperature: Omgivande |
MDL Number:
MFCD00052938 CAS nummer: 60811-23-6 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...