2-Chloro-6-fluoroaniline 98+%
Leverantör: Apollo Scientific
|
Varning
|
PC1819B-100G
PC1819B-25G
APOSPC1819B-100GEA
3125
SEK
APOSPC1819B-100G
APOSPC1819B-25G
2-Chloro-6-fluoroaniline 98+%
2-Chloro-6-fluoroaniline
Formel:
C₆H₅ClFN MW: 145,56 g/mol Kokpunkt: 67…69 °C/14mmHg Smältpunkt: 32 °C Densitet: 1,316 g/cm³ (25 °C) Flampunkt: >230 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00040309 CAS nummer: 363-51-9 EINECS: 206-657-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...