2-Fluoro-5-(trifluoromethyl)phenyl isocyanate 97%
Leverantör: Apollo Scientific
Varning
Synonyms: 2-Fluoro-5-(trifluoromethyl)phenylisocyanate
PC1789-1G PC1789-25G PC1789-5G
APOSPC1789-1GEA 855 SEK
APOSPC1789-1G APOSPC1789-25G APOSPC1789-5G
2-Fluoro-5-(trifluoromethyl)phenyl isocyanate 97%
2-Fluoro-5-(trifluoromethyl)phenyl isocyanate
  • May form small amount of sediment.
Formel: C₈H₃F₄NO
MW: 205,11 g/mol
Kokpunkt: 53 °C/2,5mmHg
Densitet: 1,418 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00673069
CAS nummer: 69922-27-6