2-Chloro-2,2-difluoroacetamide 99%

Leverantör: Apollo Scientific

PC1730-25G PC1730-5G
APOSPC1730-25GEA 2750 SEK
APOSPC1730-25G APOSPC1730-5G
2-Chloro-2,2-difluoroacetamide 99%
2-Chloro-2,2-difluoroacetamide
Formel: ClCF₂CONH₂
MW: 129,49 g/mol
Kokpunkt: 93 °C/18mmHg
Smältpunkt: 78 °C
Storage Temperature: Omgivande
MDL Number: MFCD00039775
CAS nummer: 354-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR