Potassium-3-(methoxycarbonyl)-5-nitrophenyltrifluoroborate 96%

Leverantör: Apollo Scientific

PC1709-1G PC1709-25G PC1709-5G
APOSPC1709-1GEA 1100 SEK
APOSPC1709-1G APOSPC1709-25G APOSPC1709-5G
Potassium-3-(methoxycarbonyl)-5-nitrophenyltrifluoroborate 96%
Potassium-3-(methoxycarbonyl)-5-nitrophenyltrifluoroborate
Formel: C₈H₆BF₃KNO₄
Smältpunkt: 280…284 °C
Storage Temperature: Omgivande
MDL Number: MFCD04115749
CAS nummer: 850623-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR