5-(4-Chlorobenzyloxycarbonyl)-4-(trifluoromethyl)pyrimidin-2-yl hydrazine 95+%
Leverantör: Apollo Scientific
Varning
Synonyms: 4-Chlorobenzyl 2-hydrazino-4-(trifluoromethyl)pyrimidine-5-carboxylate
PC1698-1G PC1698-250MG
APOSPC1698-1GEA 2925 SEK
APOSPC1698-1G APOSPC1698-250MG
5-(4-Chlorobenzyloxycarbonyl)-4-(trifluoromethyl)pyrimidin-2-yl hydrazine 95+%
5-(4-Chlorobenzyloxycarbonyl)-4-(trifluoromethyl)pyrimidin-2-yl hydrazine
Formel: C₁₃H₁₀ClF₃N₄O₂
MW: 346,7 g/mol
Smältpunkt: 111…113 °C
Storage Temperature: Omgivande
MDL Number: MFCD00068153
CAS nummer: 175137-33-4