5-Fluoro-(2-methoxycarbonyl)phenylboronic acid 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 5-Fluoro-2-methoxycarboxyphenylboronic acid, 5-Fluoro-2-(methoxycarbonyl)benzeneboronic acid, 2-Methoxycarbonyl-5-fluorophenylboronic acid
PC1662-1G PC1662-5G
APOSPC1662-1GEA 1775 SEK
APOSPC1662-1G APOSPC1662-5G
5-Fluoro-(2-methoxycarbonyl)phenylboronic acid 98%
5-Fluoro-(2-methoxycarbonyl)phenylboronic acid
Formel: C₈H₈BFO₄
MW: 197,96 g/mol
Smältpunkt: 146…148 °C
Storage Temperature: Kylskåp
MDL Number: MFCD03095367
CAS nummer: 850568-05-7