4-(4-Fluorophenylcarbamoyl)phenylboronic acid 97%

Leverantör: Apollo Scientific

Synonyms: (4-((4-Fluorophenyl)carbamoyl)phenyl)boronic acid, 4-[(4-Fluorophenyl)aminocarbonyl]benzeneboronic acid

PC1601-1G PC1601-25G PC1601-5G
APOSPC1601-1GEA 1800 SEK
APOSPC1601-1G APOSPC1601-25G APOSPC1601-5G
4-(4-Fluorophenylcarbamoyl)phenylboronic acid 97%
4-(4-Fluorophenylcarbamoyl)phenylboronic acid
Formel: C₁₃H₁₁BFNO₃
MW: 259,0400 g/mol
Smältpunkt: 234…240 °C
Storage Temperature: Omgivande
MDL Number: MFCD06659878
CAS nummer: 850568-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR