Bromopentafluoroethane 97%
Leverantör: Apollo Scientific
|
Varning
|
PC1530-25G
PC1530-5G
PC1530-10G
APOSPC1530-25GEA
19525
SEK
APOSPC1530-25G
APOSPC1530-5G
APOSPC1530-10G
Bromopentafluoroethane 97%
Bromopentafluoroethane
Formel:
BrF₂CCF₃ Kokpunkt: –21 °C Densitet: 1,8098 g/cm³ (25 °C) Storage Temperature: Omgivande |
CAS nummer:
354-55-2 UN: 3163 ADR: 2.2, |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...