1H,1H-Perfluoro-3,7-dimethyl-1-octanol 97%
Leverantör: Apollo Scientific
Varning
PC1218-1G PC1218-25G PC1218-5G
APOSPC1218-1GEA 860 SEK
APOSPC1218-1G APOSPC1218-25G APOSPC1218-5G
1H,1H-Perfluoro-3,7-dimethyl-1-octanol 97%
1H,1H-Perfluoro-3,7-dimethyl-1-octanol
  • Liquid at room temperature
Formel: C₁₀H₃F₁₉O
Kokpunkt: 90 °C
Storage Temperature: Omgivande
MDL Number: MFCD01320739
CAS nummer: 232587-50-7