4-Amino-α,α,α-trifluoro-m-toluic acid 98%
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
4-Amino-3-(trifluoromethyl)benzoic acid
PC1207-25G
PC1207-100G
APOSPC1207-25GEA
2150
SEK
APOSPC1207-25G
APOSPC1207-100G
4-Amino-α,α,α-trifluoro-m-toluic acid 98%
4-Amino-α,α,α-trifluoro-m-toluic acid
Formel:
C₈H₆F₃NO₂ MW: 205,14 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD03407960 CAS nummer: 400-76-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...