Potassium-4-(methoxycarbonyl)phenyltrifluoroborate

Leverantör: Apollo Scientific
Varning

Synonyms: Potassium trifluoro(4-(methoxycarbonyl)phenyl)borate

PC11240-1G PC11240-5G
APOSPC11240-1GEA 1075 SEK
APOSPC11240-1G APOSPC11240-5G
Potassium-4-(methoxycarbonyl)phenyltrifluoroborate
Potassium-4-(methoxycarbonyl)phenyltrifluoroborate
Formel: C₈H₇BF₃KO₂
MW: 242,05 g/mol
Smältpunkt: >300 °C
Storage Temperature: Omgivande
MDL Number: MFCD04115741
CAS nummer: 705254-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR