Potassium (3-cyanophenyl)trifluoroborate
Leverantör: Apollo Scientific
Synonyms:
(3-Cyanophenyl)trifluoroboric acid potassium salt
PC11239-25G
PC11239-5G
APOSPC11239-25GEA
5475
SEK
APOSPC11239-25G
APOSPC11239-5G
Potassium (3-cyanophenyl)trifluoroborate
Potassium (3-cyanophenyl)trifluoroborate
Formel:
C₇H₄BF₃KN Smältpunkt: 196…200 °C Storage Temperature: Omgivande |
MDL Number:
MFCD04115744 CAS nummer: 850623-46-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...