3-Amino-2,6-dichloro-4-(trifluoromethyl)pyridine 98%

Leverantör: Apollo Scientific
Varning

Synonyms: 2,6-Dichloro-4-(trifluoromethyl)pyridin-3-amine

PC1070O-10G PC1070O-1G
APOSPC1070O-10GEA 3900 SEK
APOSPC1070O-10G APOSPC1070O-1G
3-Amino-2,6-dichloro-4-(trifluoromethyl)pyridine 98%
3-Amino-2,6-dichloro-4-(trifluoromethyl)pyridine
Formel: C₆H₃Cl₂F₃N₂
MW: 231,01 g/mol
Smältpunkt: 45…48 °C
Storage Temperature: Omgivande
MDL Number: MFCD00173957
CAS nummer: 175277-67-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR