Zinc(II) Bis(trifluoromethanesulphonyl)imide
Leverantör: Apollo Scientific
Varning
Synonyms: Zinc bis(trifluoromethylsulphonyl)imide, Bis(trifluoromethanesulphonyl)imide Zinc(II) Salt, Zinc(II) Triflimide
PC10697-1G PC10697-250MG PC10697-5G PC10697-25G
APOSPC10697-1GEA 775 SEK
APOSPC10697-1G APOSPC10697-250MG APOSPC10697-5G APOSPC10697-25G
Zinc(II) Bis(trifluoromethanesulphonyl)imide
Zinc(II) Bis(trifluoromethanesulphonyl)imide
Formel: C₄F₁₂N₂O₈S₄Zn
MW: 625,65 g/mol
Smältpunkt: 143…147 °C
Storage Temperature: Omgivande
MDL Number: MFCD16621474
CAS nummer: 168106-25-0