2-Chloro-3-nitro-5-(trifluoromethyl)pyridine 98%
Leverantör: Apollo Scientific
Varning
PC10641-5G PC10641-25G
APOSPC10641-5GEA 325 SEK
APOSPC10641-5G APOSPC10641-25G
2-Chloro-3-nitro-5-(trifluoromethyl)pyridine 98%
2-Chloro-3-nitro-5-(trifluoromethyl)pyridine
Formel: C₆H₂ClF₃N₂O₂
Storage Temperature: Kylskåp
MDL Number: MFCD06255171
CAS nummer: 72587-15-6
UN: 2810
ADR: 6.1,III