2,6-Bis(trifluoromethyl)phenylboronic acid
Leverantör: Apollo Scientific
Varning
Synonyms: 2,6-Bis(trifluoromethyl)benzeneboronic acid
PC10506-1G PC10506-250MG PC10506-5G
APOSPC10506-1GEA 1750 SEK
APOSPC10506-1G APOSPC10506-250MG APOSPC10506-5G
2,6-Bis(trifluoromethyl)phenylboronic acid
2,6-Bis(trifluoromethyl)phenylboronic acid
Formel: C₈H₅BF₆O₂
MW: 257,93 g/mol
Smältpunkt: ∼ 150 °C
Storage Temperature: Frys
MDL Number: MFCD04039321
CAS nummer: 681812-07-7