2-Fluoro-4-methylphenylboronic acid
Leverantör: Apollo Scientific
Varning
Synonyms: 2-Fluoro-4-methylbenzeneboronic acid, (2-Fluoro-4-methylphenyl)boronic acid
PC10336-1G PC10336-25G PC10336-5G
APOSPC10336-1GEA 175 SEK
APOSPC10336-1G APOSPC10336-25G APOSPC10336-5G
2-Fluoro-4-methylphenylboronic acid
2-Fluoro-4-methylphenylboronic acid
Formel: C₇H₈BFO₂
MW: 153,95 g/mol
Smältpunkt: 240…245 °C
Storage Temperature: Omgivande
MDL Number: MFCD05664239
CAS nummer: 170981-26-7