2-[(3,3,3-Trifluoro-2-hydroxypropyl)sulphanyl]benzenecarboxylic acid
Leverantör: Apollo Scientific
Varning
Synonyms: 2-[(2-Hydroxy-3,3,3-trifluoroprop-1-yl)thio]benzoic acid
PC10296-1G PC10296-500MG
APOSPC10296-1GEA 2150 SEK
APOSPC10296-1G APOSPC10296-500MG
2-[(3,3,3-Trifluoro-2-hydroxypropyl)sulphanyl]benzenecarboxylic acid
2-[(3,3,3-Trifluoro-2-hydroxypropyl)sulphanyl]benzenecarboxylic acid
Formel: C₁₀H₉F₃O₃S
MW: 266,24 g/mol
Smältpunkt: 98…100 °C
Storage Temperature: Omgivande
MDL Number: MFCD05663825
CAS nummer: 886361-21-3