3-(4-Chlorophenyl)-3-[(2,2,2-trifluoroacetyl)amino]propanoic acid
Leverantör: Apollo Scientific
Varning
Synonyms: 3-(4-Chlorophenyl)-3-(trifluoroacetamido)propanoic acid
PC10222-10G PC10222-1G PC10222-5G
APOSPC10222-10GEA 9425 SEK
APOSPC10222-10G APOSPC10222-1G APOSPC10222-5G
3-(4-Chlorophenyl)-3-[(2,2,2-trifluoroacetyl)amino]propanoic acid
3-(4-Chlorophenyl)-3-[(2,2,2-trifluoroacetyl)amino]propanoic acid
Formel: C₁₁H₉ClF₃NO₃
MW: 295,65 g/mol
Smältpunkt: 162…165 °C
Storage Temperature: Omgivande
MDL Number: MFCD03791205
CAS nummer: 117291-25-5