1H,1H,8H-Perfluoro-1-octanol
Leverantör: Apollo Scientific
Varning
Synonyms: 1H,1H,8H-Tetradecafluorooctan-1-ol
PC0935-1G PC0935-5G
APOSPC0935-1GEA 1125 SEK
APOSPC0935-1G APOSPC0935-5G
1H,1H,8H-Perfluoro-1-octanol
1H,1H,8H-Perfluoro-1-octanol
Formel: C₈H₄F₁₄O
MW: 382,1 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD01631687
CAS nummer: 10331-08-5