4-Fluorophenyl acetate 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 4-Fluorophenyl Acetate, 1-Acetoxy-4-fluorobenzene, Acetic acid 4-Fluorophenyl acetate, 1-Acetoxy-4-fluorobenzene
PC0910-100G PC0910-25G PC0910-5G
APOSPC0910-100GEA 1550 SEK
APOSPC0910-100G APOSPC0910-25G APOSPC0910-5G
4-Fluorophenyl acetate 98%
4-Fluorophenyl acetate
Formel: C₈H₇FO₂
Kokpunkt: 197…199 °C
Densitet: 1,178 g/cm³ (25 °C)
Flampunkt: 87 °C
Storage Temperature: Omgivande
MDL Number: MFCD00041361
CAS nummer: 405-51-6
EINECS: 609-847-6