Fmoc-3-(trifluoromethyl)-D-phenylalanine 99+%
Leverantör: Apollo Scientific
Synonyms: 3-(Trifluoromethyl)-D-phenylalanine, N-FMOC protected, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3-(trifluoromethyl)phenyl)propanoic acid, Fmoc-D-Phe(3-CF₃)-OH
PC0812-1G PC0812-5G
APOSPC0812-1GEA 3600 SEK
APOSPC0812-1G APOSPC0812-5G
Fmoc-3-(trifluoromethyl)-D-phenylalanine 99+%
Fmoc-3-(trifluoromethyl)-D-phenylalanine
Formel: C₂₅H₂₀F₃NO₄
MW: 455,44 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00672556
CAS nummer: 205526-28-9