3-(Pentafluorothio)benzaldehyde 97%
Leverantör: Apollo Scientific
|
Varning
|
PC0796-1G
PC0796-5G
APOSPC0796-1GEA
5075
SEK
APOSPC0796-1G
APOSPC0796-5G
3-(Pentafluorothio)benzaldehyde 97%
3-(Pentafluorothio)benzaldehyde
Formel:
C₇H₅F₅OS MW: 232,17 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD03788517 CAS nummer: 401892-80-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...