Tris(pentafluorophenyl)borane 97%
Leverantör: Apollo Scientific
Varning
Synonyms: Tris(perfluorophenyl)borane
PC0791-1G PC0791-5G
APOSPC0791-1GEA 345 SEK
APOSPC0791-1G APOSPC0791-5G
Tris(pentafluorophenyl)borane 97%
Tris(pentafluorophenyl)borane
Useful component of Ziegler-Nattametallocene catalyst for polyolefins.J. Organomet. Chem., 1964, 2, 245
Formel: C₁₈BF₁₅
MW: 511.98 g/mol
Smältpunkt: 126…131 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00269813
CAS nummer: 1109-15-5