3-(Pentafluorothio)aniline 97%

Leverantör: Apollo Scientific
Fara

Synonyms: 3-Aminophenylsulphurpentafluoride, 3-Aminophenylsulphur pentafluoride

PC0779-1G PC0779-25G PC0779-5G
APOSPC0779-1GEA 3350 SEK
APOSPC0779-1G APOSPC0779-25G APOSPC0779-5G
3-(Pentafluorothio)aniline 97%
3-(Pentafluorothio)aniline
Formel: C₆H₆F₅NS
MW: 218,17 g/mol
Kokpunkt: 87 °C/3,2mmHg
Smältpunkt: 35 °C
Storage Temperature: Omgivande
MDL Number: MFCD00674199
CAS nummer: 2993-22-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR