2-(Trifluoromethyl)phenylboronic acid 98%
Leverantör: Apollo Scientific
Varning
Synonyms: (2-Trifluoromethyl)phenylboronic acid, (2-(Trifluoromethyl)phenyl)metaboric acid, Dihydroxy(2-(trifluoromethyl)phenyl)borane, 2-(Trifluoromethyl)benzeneboronic acid
PC0677-100G PC0677-25G PC0677-5G
APOSPC0677-100GEA 1550 SEK
APOSPC0677-100G APOSPC0677-25G APOSPC0677-5G
2-(Trifluoromethyl)phenylboronic acid 98%
2-(Trifluoromethyl)phenylboronic acid
Formel: C₇H₆BF₃O₂
MW: 189.93 g/mol
Smältpunkt: 108…114 °C
Storage Temperature: Omgivande
MDL Number: MFCD00236059
CAS nummer: 1423-27-4