(R)-(-)-1,1'-Binaphthyl-2,2'-diyl bis(trifluoromethanesulphonate) 98%
Leverantör: Apollo Scientific
Fara
Synonyms: (R)-(-)-1,1'-Binaphthol-2,2'-bis(trifluoromethanesulphonate), (R)-(-)-1,1'-Bi-2-naphthol Bis(trifluoromethanesulphonate), (R)-(-)-1,1'-Bi-2-naphthol bis(trifluoromethanesulphonate), (R)-[1,1'-Binaphthalene]-2,2'-diyl bis(trifluoromethanesulphonate)
PC0614-1G PC0614-5G PC0614-25G
APOSPC0614-1GEA 720 SEK
APOSPC0614-1G APOSPC0614-5G APOSPC0614-25G
(R)-(-)-1,1'-Binaphthyl-2,2'-diyl bis(trifluoromethanesulphonate) 98%
(R)-(-)-1,1'-Binaphthyl-2,2'-diyl bis(trifluoromethanesulphonate)
Formel: C₂₂H₁₂F₆O₆S₂
MW: 550,44 g/mol
Smältpunkt: 80…85 °C
Storage Temperature: Omgivande
MDL Number: MFCD00274615
CAS nummer: 126613-06-7
UN: 3261
ADR: 8,II