α,α,α,3-Tetrafluoro-o-toluic acid 97%
Leverantör: Apollo Scientific
Varning
Synonyms: 3-Fluoro-2-trifluoromethylbenzoic acid, 3-Fluoro-2-(trifluoromethyl)benzoic acid
PC0388-1G PC0388-5G PC0388-25G
APOSPC0388-1GEA 380 SEK
APOSPC0388-1G APOSPC0388-5G APOSPC0388-25G
α,α,α,3-Tetrafluoro-o-toluic acid 97%
α,α,α,3-Tetrafluoro-o-toluic acid
Formel: C₈H₄F₄O₂
MW: 208,11 g/mol
Smältpunkt: 119…121 °C
Storage Temperature: Omgivande
MDL Number: MFCD01631565
CAS nummer: 261951-80-8