3-(3,4-Difluorophenyl)propionic acid 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 3-(3,4-Difluorophenyl)propanoic acid
PC0334-25G PC0334-5G PC0334-1G
APOSPC0334-25GEA 3100 SEK
APOSPC0334-25G APOSPC0334-5G APOSPC0334-1G
3-(3,4-Difluorophenyl)propionic acid 98%
3-(3,4-Difluorophenyl)propionic acid
Formel: C₉H₈F₂O₂
MW: 186,16 g/mol
Smältpunkt: 49…53 °C
Storage Temperature: Omgivande
MDL Number: MFCD00799520
CAS nummer: 161712-75-0