2,4-Difluoro-5-(trifluoromethyl)aniline
Leverantör: Apollo Scientific
Varning
Synonyms: 5-Amino-2,4-difluorobenzotrifluoride
PC0286-1G PC0286-250MG
APOSPC0286-1GEA 5475 SEK
APOSPC0286-1G APOSPC0286-250MG
2,4-Difluoro-5-(trifluoromethyl)aniline
2,4-Difluoro-5-(trifluoromethyl)aniline
Formel: C₇H₄F₅N
MW: 197,1054 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD01631613
CAS nummer: 261944-56-3