N-metyl-N-(trimetylsilyl) trifluoracetamid 98%
Leverantör: Apollo Scientific
Varning
Synonyms: MSTFA, N-Methyl-N-(trimethylsilyl)-2,2,2-trifluoroacetamide, 2,2,2-Trifluoro-N-methyl-N-(trimethylsilyl)acetamide
PC0132-250G PC0132-25G PC0132-50G
APOSPC0132-250GEA 2075 SEK
APOSPC0132-250G APOSPC0132-25G APOSPC0132-50G
N-metyl-N-(trimetylsilyl) trifluoracetamid 98%
N-Methyl-N-(trimethylsilyl)trifluoroacetamide
A powerful silylation reagent with volatile byproducts:J. Chromat., 115, 591 (1975); 134, 387 (1977).
Formel: C₆H₁₂F₃NOSi
MW: 199,25 g/mol
Kokpunkt: 130…132 °C
Densitet: 1,074 g/cm³ (25 °C)
Flampunkt: 25 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00000411
CAS nummer: 24589-78-4
EINECS: 246-331-6
UN: 1993
ADR: 3,III