Perfluorocyclohexene

Leverantör: Apollo Scientific
Varning

PC0101-25G PC0101-5G
APOSPC0101-25GEA 5475 SEK
APOSPC0101-25G APOSPC0101-5G
Perfluorocyclohexene
Perfluorocyclohexene
Formel: C₆F₁₀
Kokpunkt: 52…52,5 °C
Smältpunkt: –50 °C
Densitet: 1,665 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00037153
CAS nummer: 355-75-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR