4-(2-Aminoethyl)benzenesulphonamide

Leverantör: Apollo Scientific
Fara

OR9989-100G OR9989-25G OR9989-500G
APOSOR9989-100GEA 685 SEK
APOSOR9989-100G APOSOR9989-25G APOSOR9989-500G
4-(2-Aminoethyl)benzenesulphonamide
4-(2-Aminoethyl)benzenesulphonamide
Formel: C₈H₁₂N₂O₂S
Smältpunkt: 150…152 °C
Storage Temperature: Omgivande
MDL Number: MFCD00010301
CAS nummer: 35303-76-5
EINECS: 252-501-0
UN: 3259
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR