6-Nitro-m-toluic acid
Leverantör: Apollo Scientific
Varning
Synonyms: 2-Nitro-5-methylbenzoic acid, 5-Methyl-2-nitrobenzoic acid
OR9984-10G OR9984-25G
APOSOR9984-10GEA 175 SEK
APOSOR9984-10G APOSOR9984-25G
6-Nitro-m-toluic acid
6-Nitro-m-toluic acid
Formel: C₈H₇NO₄
MW: 181,15 g/mol
Smältpunkt: 133…136 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007368
CAS nummer: 3113-72-2