3-(tert-Butyl)-2,4-pentanedione, Technical Grade
Leverantör: Apollo Scientific
Varning
OR9919-1G OR9919-500MG
APOSOR9919-1GEA 3875 SEK
APOSOR9919-1G APOSOR9919-500MG
3-(tert-Butyl)-2,4-pentanedione, Technical Grade
3-(tert-Butyl)-2,4-pentanedione
Formel: C₉H₁₆O₂
MW: 156,23 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD01846256
CAS nummer: 13221-94-8