2-Amino-1-phenylethanol, Technical Grade
Leverantör: Apollo Scientific
Fara
Synonyms: alpha-(Aminomethyl)benzyl Alcohol, DL-β-Hydroxyphenethylamine, Phenylethanolamine
OR9824-100G OR9824-25G OR9824-500G
APOSOR9824-100GEA 945 SEK
APOSOR9824-100G APOSOR9824-25G APOSOR9824-500G
2-Amino-1-phenylethanol, Technical Grade
2-Amino-1-phenylethanol
Formel: C₈H₁₁NO
MW: 137,18 g/mol
Kokpunkt: 160 °C/17mmHg
Smältpunkt: 56…58 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008137
CAS nummer: 7568-93-6
EINECS: 231-469-1
UN: 3259
ADR: 8,III