3-Pyrid-4-ylbenzoic acid 95%
Leverantör: Apollo Scientific
Varning
Synonyms: 3-(Pyridin-4-yl)benzoic acid
OR9726-1G OR9726-250MG
APOSOR9726-1GEA 1150 SEK
APOSOR9726-1G APOSOR9726-250MG
3-Pyrid-4-ylbenzoic acid 95%
3-Pyrid-4-ylbenzoic acid
Formel: C₁₂H₉NO₂
MW: 199,2054 g/mol
Smältpunkt: 268 °C
Storage Temperature: Omgivande
MDL Number: MFCD05864808
CAS nummer: 4385-78-8