2-Bromo-4-chlorotoluene

Leverantör: Apollo Scientific
Varning

OR9677-100G OR9677-25G
APOSOR9677-100GEA 650 SEK
APOSOR9677-100G APOSOR9677-25G
2-Bromo-4-chlorotoluene
2-Bromo-4-chlorotoluene
Formel: C₇H₆BrCl
Kokpunkt: 114…12mm °C
Densitet: 156 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00060650
CAS nummer: 27139-97-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR