4-(6-Methylpyrazin-2-yl)-1,4-diazepane 97%
Leverantör: Apollo Scientific
Synonyms: 1-(6-Methylpyrazin-2-yl)homopiperazine
OR9662-1G OR9662-250MG
APOSOR9662-1GEA 4875 SEK
APOSOR9662-1G APOSOR9662-250MG
4-(6-Methylpyrazin-2-yl)-1,4-diazepane 97%
4-(6-Methylpyrazin-2-yl)-1,4-diazepane
Formel: C₁₀H₁₆N₄
MW: 192.27 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD08435920
CAS nummer: 869901-23-5