4,6-Dichloroquinazoline 95%
Leverantör: Apollo Scientific
OR965616-1G
OR965616-250MG
APOSOR965616-1GEA
3175
SEK
APOSOR965616-1G
APOSOR965616-250MG
4,6-Dichloroquinazoline 95%
4,6-Dichloroquinazoline
Formel:
C₈H₄Cl₂N₂ MW: 199,0368 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD00089492 CAS nummer: 7253-22-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...