(±)-2-Metylhexansyra 95%

Leverantör: Apollo Scientific

Synonyms: 2-MethylhexanoicAcid, 2-Methylhexanoic acid, (±)-2-Methylhexanoic acid

OR965380-100G OR965380-25G
APOSOR965380-100GEA 3175 SEK
APOSOR965380-100G APOSOR965380-25G
(±)-2-Metylhexansyra 95%
(±)-2-Metylhexansyra
Formel: C₇H₁₄O₂
MW: 130,19 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00002674
CAS nummer: 4536-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR