1-(3-(Chloromethyl)phenyl)ethanone 95%

Leverantör: Apollo Scientific

Synonyms: 3'-Chloromethylacetophenone

OR964995-1G OR964995-250MG
APOSOR964995-1GEA 5150 SEK
APOSOR964995-1G APOSOR964995-250MG
1-(3-(Chloromethyl)phenyl)ethanone 95%
1-(3-(Chloromethyl)phenyl)ethanone
Formel: C₉H₉ClO
MW: 168,6202 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD06658170
CAS nummer: 41908-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR