3-Aminophthalic acid, Technical Grade
Leverantör: Apollo Scientific
Varning
OR964780-1G OR964780-25G OR964780-5G OR964780-10G
APOSOR964780-1GEA 380 SEK
APOSOR964780-1G APOSOR964780-25G APOSOR964780-5G APOSOR964780-10G
3-Aminophthalic acid, Technical Grade
3-Aminophthalic acid
Formel: C₈H₇NO₄
MW: 181,15 g/mol
Smältpunkt: 183 °C
Storage Temperature: Kylskåp
CAS nummer: 5434-20-8