2-(4-Chlorophenyl)-2-propanol 98%
Leverantör: Apollo Scientific
Synonyms: 4-Chloro-alpha,alpha-dimethylbenzyl alcohol
OR962886-1G OR962886-250MG OR962886-5G
APOSOR962886-1GEA 2750 SEK
APOSOR962886-1G APOSOR962886-250MG APOSOR962886-5G
2-(4-Chlorophenyl)-2-propanol 98%
2-(4-Chlorophenyl)-2-propanol
Formel: C₉H₁₁ClO
MW: 170,64 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11870102
CAS nummer: 1989-25-9